missing translation for 'onlineSavingsMsg'
Learn More

Fluoride Standard for Ion Chromatography, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

£40.00 - £110.84

Chemical Identifiers

CAS 7732-18-5
View More Specs

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10288772
View Documents Promotion Details
SPEX Certiprep
AS-F9-2Y
125 mL Triple-leached LDPE bottle
£108.00
125mL
Please to purchase this item. Need a web account? Register with us today!
11480269
View Documents
SPEX Certiprep
AS-F9-2Y
125 mL Triple-leached LDPE bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11466108
View Documents
SPEX Certiprep
AS-F9-2X
500 mL Triple-leached LDPE bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
F-1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Identifiers

7732-18-5

Specifications

7732-18-5
0.1, 99.9
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
1000μg/mL F−
Fluoride Standard
0.1, 99.9
With Certificate of Analysis.
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
H2O
Videos
Safety and Handling

Safety and Handling

missing translation for 'dotInformation' : Nonhazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Product Certifications

Product intended for laboratory use only