missing translation for 'onlineSavingsMsg'
Learn More

Ion Chromatography (IC) Multi Element Anion Standard in Water (5 Elements), Reagecon
BIOPHARMA

Manufactued from ultra-pure salts gravimetrically

Brand:  Reagecon ICAS501

Product Code. 11526693

  • £223.00 / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Tested by ISO 17025 accreditation for calibration of laboratory balances
  • ISO/IEC 17025 for:
    • Temperature calibration in -90 to 650 °C
    • Density measurement using oscillating u-tube and Bingham pycnometry methods
    • Certain complexometric titrations
  • All volumetric titrimetric and gravimetric functions carried out under highly regulated temp regime
  • Produced in highly controlled cleanroom environment using ultra-pure water, triple distilled acids and triple leached pre-cleaned bottles
  • All uncertainties of measurement calculated according to Eurachem/CITAC guidelines and reported at 95% confidence level
Specifications

Specifications

HDPE triple-leached
Ion Chromatography Standard
Water
IC and other Anion Measurement Techniques
100 mL
100 μg/mL each of fluoride and chloride, 200 μg/mL each of nitrate, phosphate and sulphate
GHS/OSHA compliant, REACH
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers