Learn More
Instrument Check Standard 2 For Ion Chromatography , SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep ICMIX2-100
Code : NEW
Additional Details : Weight : 0.12500kg
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
HPO4-2 | 600μg/mL |
Br- | 400μg/mL |
NO3- | 400μg/mL |
SO4 | 400μg/mL |
Cl- | 200μg/mL |
F- | 100μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
IC Instrument Check Standard 2 | |
125 mL | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Components in H2O: Br− •400μg/mL Cl− •200μg/mL F− •100μg/mL NO3− •400μg/mL HPO4−2 •600μg/mL SO4−2 •400μg/mL |
Triple-leached LDPE bottle | |
With Certificate of Analysis | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
H2O |
Product intended for laboratory use only