missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Instrument Check Standard 6 For Ion Chromatography, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep ICMIX6-100
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
HPO4-2 | 150μg/mL |
SO4-2 | 150μg/mL |
Br- | 100μg/mL |
Cl- | 50μg/mL |
NO2-N | 25μg/mL |
NO3-N | 25μg/mL |
F- | 20μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Instrument Check Standard 6 | |
125 mL | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Components in water: Br- •100μg/mL Cl- •50μg/mL F- •20μg/mL HPO4-2 •150μg/mL NO3--N •25μg/mL NO2--N •25μg/mL SO4-2 •150μg/mL |
Triple-leached LDPE bottle | |
With Certificate of Analysis | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
H2O |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Product intended for laboratory use only
Spot an opportunity for improvement?Share a Content Correction