missing translation for 'onlineSavingsMsg'
Learn More

Phosphate-P Standard for Ion Chromatography, SPEX CertiPrep™

Catalog Number 15648235
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Triple-leached LDPE bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
PO&subscript_4;-P1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Packaging Triple-leached LDPE bottle
CAS Min % 0.1, 99.9
Quantity 125 mL
CAS Max % 0.1, 99.9
CAS 7732-18-5
Certificate of Analysis With Certificate of Analysis
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Concentration or Composition (by Analyte or Components) 10002g/mL
Solvent or Matrix H2O
Chemical Name or Material Phosphate-Phosphorus Standard
Show More Show Less

Product intended for laboratory use only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'