missing translation for 'onlineSavingsMsg'
Learn More

TaqMan™ Ion Torrent™ Ion Library TaqMan™ Quantitation Kit

An easy-to-use, flexible solution for the accurate quantitation of library input for the Ion Torrent semiconductor sequencing workflow

Brand:  TaqMan™ Ion Torrent™ 4468802

1556.10 GBP valid until 2025-12-31
Use promo code "24365" to get your promotional price.


Product Code. 15501474

  • £1718.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Precision: The TaqMan™ Assay provides specificity and accuracy in detecting properly adapted, amplifiable templates over non-probe-based quantitation methods
  • Sensitivity: The protocol supports quantitation of femtomolar amounts of fragment libraries
  • Simplicity: The validated, ready-to-use qPCR standard requires only a simple serial dilution to determine the optimal concentration of library to use for templated bead preparation
  • Versatility: The highly robust qPCR mix accommodates a wide range of cycling conditions and reaction volumes, and combines highly sensitive detection with a broad quantification range
  • Efficiency: qPCR-based methods use less material for quantification
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sequencing - Library Quantitation
Kit
Visible
Sequencing
Ion Torrent™
250 reactions, 2 Tubes
Wet Ice
Store components at -20° C
Visible
Ion PGM™ System and Ion S5 Plus
250
Quantitation Kit
2.5 ml
Library Quantitation
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2864360 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number4468802
Lot Number2758172 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number4468802
Lot Number2758173 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number4468802
Lot Number2809033 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number4468802
Lot Number2809114 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number4468802
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
TaqMan™ Ion Torrent™ Ion Library TaqMan™ Quantitation Kit > 250 Reactions

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.