Learn More
Sartorius™ Ionizing Blower
Neutralize static electricity within seconds. Sartorius™ Ionizing Blower works by shooting ions at the material to be weighed.
Brand:  Sartorius™ YIB01-0DR
Product Code. 10459312
  • £1750.00 / Each
Explore more special offers
Description
Description
  • Neutralizes static electricity
  • Eliminates the need for complicated, expensive, and often ineffective humidity change approaches.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
230 V, 50 Hz
0.6 kg