missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ionomycin Calcium Salt, 99%

Product Code. 15433369
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 15433369

Brand: Thermo Scientific Chemicals J60628.#0

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 56092-82-1 | C41H70CaO9, C41H70CaO9 | 747.08 g/mol

Ionomycin calcium salt is used as a polyether antibiotic and apoptotic degeneration inducer. Ionomycin induces apoptotic degeneration of embryonic cortical neurons and cell cycle arrest at G1 phase. It is more effective than A23187 (sc-3591) and provides a useful tool for increasing intracellular Ca2+ concentration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mg
Description White to off-white
Content And Storage -30°C to -10°C
For Use With (Application) Highly selective and potent calcium ionophore
CAS 56092-82-1
Molecular Formula C41H70CaO9
Formula Weight 747.08
Sensitivity Light Sensitive
Solubility Soluble in DMSO (10mg/ml). Also soluble in ethanol or methanol
Product Type Ionomycin Calcium Salt, 99%
Form Crystalline powder
Beilstein 1416163
InChI Key RULWQOBWZOQCOY-SQNLDGQHSA-M
SMILES [Ca++].C[C@H](CCC([O-])=O)C[C@H](C)C[C@H](C)C(=O)[CH-]C(=O)[C@H](C)C[C@H](C)C\C=C\[C@@H](C)[C@@H](O)[C@@H](C)[C@@H](O)C[C@@H]1CC[C@](C)(O1)[C@H]1CC[C@](C)(O1)[C@@H](C)O
IUPAC Name calcium (4R,6S,8S,12R,14R,16E,18R,19R,20S,21S)-19,21-dihydroxy-22-[(2S,2'R,5S,5'S)-5'-[(1R)-1-hydroxyethyl]-2,5'-dimethyl-[2,2'-bioxolan]-5-yl]-4,6,8,12,14,18,20-heptamethyl-9,11-dioxodocos-16-en-10-idoate
Molecular Weight (g/mol) 747.08
Assay 99%
Show More Show Less
compliance-icons
Product Identifier
  • Ionomycin calcium salt
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Ionomycin calcium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.