missing translation for 'onlineSavingsMsg'
Learn More
Ionomycin calcium salt, Tocris Bioscience™ Product Code.: 30001281

Ionomycin calcium salt, Tocris Bioscience™

Product Code. 30001281
1 mg
Click to view available options
Quantity:
1 mg
This item is not returnable. View return policy

Product Code. 30001281

Brand: Tocris Bioscience 1704/1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Calcium ionophore

Ionomycin calcium salt is a calcium ionophore; more specific than A23187 .

Free Acid also available.

TRUSTED_SUSTAINABILITY

Specifications

Type Ionomycin Calcium Salt
CAS 56092-82-1
Quantity 1 mg
Molecular Formula C41H70CaO9
MDL Number MFCD00083634
Synonym (4R,6S,8S,10Z,12R,14R,16E,18R,19R,20S,21S)-11,19,21-Trihydroxy-4,6,8,12,14,18,20-heptamethyl-22-[(2S,2'R,5S,5'S)-octahydro-5'-[(1R)-1-hydroxyethyl]-2,5'-dimethyl[2,2'
InChI Key RULWQOBWZOQCOY-SQNLDGQHSA-M
SMILES [Ca++].C[C@H](CCC([O-])=O)C[C@H](C)C[C@H](C)C(=O)[CH-]C(=O)[C@H](C)C[C@H](C)C\C=C\[C@@H](C)[C@@H](O)[C@@H](C)[C@@H](O)C[C@@H]1CC[C@](C)(O1)[C@H]1CC[C@](C)(O1)[C@@H](C)O
IUPAC Name calcium (4R,6S,8S,12R,14R,16E,18R,19R,20S,21S)-19,21-dihydroxy-22-[(2S,2'R,5S,5'S)-5'-[(1R)-1-hydroxyethyl]-2,5'-dimethyl-[2,2'-bioxolan]-5-yl]-4,6,8,12,14,18,20-heptamethyl-9,11-dioxodocos-16-en-10-idoate
Molecular Weight (g/mol) 747.08
PubChem CID 71311740
Formula Weight 747.08
Percent Purity >98%
Chemical Name or Material Ionomycin calcium salt
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ionomycin calcium salt, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.