Learn More
Gibco™ IP10 Recombinant Mouse Protein
Recombinant IP10 is a bioactive protein intended for use in cell culture applications. IP10 is a chemokine of the CXC subfamily and ligand for the receptor CXCR3. Binding of this protein to CXCR3 results in pleiotropic effects, including stimulation of monocytes, natural killer and T-cell migration, and modulation of adhesion molecule expression.
Brand: Gibco™ PMC1081
Code : 50
Additional Details : Weight : 0.50000kg
Specifications
Cell Culture | |
Sequential Chromatography | |
Store in refrigerator (2–8°C) | |
IP-10 | |
Recombinant | |
Chemokines & Receptors | |
IP-10 |
8.7 kDa | |
100μg | |
< 0.1 ng/μg | |
Mouse | |
E. coli | |
Lyophilized | |
95 % |
For Research Use Only. Not for use in diagnostic procedures.