Learn More
Cytiva IPGbox
Tools for enhancing reswelling of precast Immobiline DryStrip gels (IPG strips)
Brand: Cytiva 28-9334-65
Code : GE
Additional Details : Weight : 1.00000kg
Includes: IPGbox: 1 IPGbox, 1 IPGbox Kit (10 Reswell trays, 1 IPGbox insert, instructions)
Description
- Oil free rehydratation of up to 12 Immobiline DryStrip gels or other IPG strips independently and simultaneously
- Lid protects IPG strips from dust and contamination
- Load samples for IEF during rehydration by including them in the rehydration bufferor load sample using sample cups
- IPGbox used together with IPGbox Kit (Cat. No 45002627) to rehydrate 7 to 24cm Immobiline DryStrip gels
- Includes reswelling tray made of hydrophobic material to prevent sticking of rehydration solution
- IPGbox Insert located within lid of IPGbox to maintain optimal conditions for rehydration
Specifications
IPGBox | |
1 IPGbox and 1 IPGbox kit | |
Up to 12 Immobiline dry strip gel |