Learn More
Cytiva IPGbox Kit
Tools for enhancing reswelling of precast Immobiline DryStrip gels (IPG strips)
Brand: Cytiva 28-9334-92
Code : GE
Additional Details : Weight : 1.00000kg
Includes: IPGbox Kit: 10 Reswell trays, 1 IPGbox insert, instructions
Description
- Oil free rehydratation of up to 12 Immobiline DryStrip gels or other IPG strips independently and simultaneously
- Lid protects IPG strips from dust and contamination
- Load samples for IEF during rehydration by including them in rehydration bufferor load sample using sample cups
- IPGbox (Cat. No. 45002631) used together with IPGbox Kit to rehydrate 7 to 24cm Immobiline DryStrip gels
- Includes reswelling tray made of hydrophobic material to prevent sticking of rehydration solution
- IPGbox Insert located within lid of IPGbox to maintain optimal conditions for rehydration
Specifications
IPGBox Kit | |
10 Reswell trays and 1 IPGbox insert | |
PET |