Learn More
Invitrogen™ iPrep™ Card: gDNA Blood
iPrep™ Card: gDNA Blood
Brand: Invitrogen™ IS10012
Code : 50
Additional Details : Weight : 0.44000kg
Description
The iPrep™ Protocol Card is a flash, 512 KB memory card. Each iPrep™ Protocol card is pre-programmed with the purification protocol that directs the volume of reagents used and incubation time.The following iPrep™ Protocol Cards are available from Invitrogen:
- iPrep™ Forensic Card for use with the iPrep™ ChargeSwitch™ Forensic Kit and the iPRep™ ChageSwitch™ gDNA Buccal kit
-iPrep™ Tissue Card for use with the iPrep ChargeSwitch™ Tissue Kit
- iPrep™ Blood Card for use with the iPrep PureLink Blood kit
Specifications
Store at room temperature. | |
iPrep™ | |
iPrep™ | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.