Learn More
Invitrogen™ iPrep™ Card: Viral DNA/RNA
iPrep™ Card: Viral DNA/RNA
Brand:  Invitrogen™ IS10016
Product Code. 10317654
  • £596.00 / Each
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Description
Description
The iPrep™ Viral DNA/RNA protocol Card is for use with the iPrep™ PureLink™ Virus Kit. The iPrep™ Protocol Card is a flash, 512 KB memory card. Each iPrep™ Protocol card is pre-programmed with the purification protocol that directs the volume of reagents used and incubation time. The following iPrep™ Protocol Cards are available from Invitrogen:
  • iPrep™ Forensic Card for use with the iPrep™ ChargeSwitch™ Forensic Kit and the iPRep™ ChageSwitch™ gDNA Buccal kit
  • iPrep™ Tissue Card for use with the iPrep™ ChargeSwitch™ Tissue Kit
  • iPrep™ Blood Card for use with the iPrep™ PureLink™ Blood kit
  • iPrep™ Viral DNA/RNA Card for use with the iPrep™ PureLink™ Virus Kit
    TRUSTED_SUSTAINABILITY
  • Specifications
    Specifications
    Card
    iPrep™
    1 card
    Product Suggestions
    Product Suggestions
    Videos
    SDS
    Documents
    Documents
    One moment while we fetch your results.
    Certificates
    Special Offers
    Special Offers
    Product Content Correction
    Product Title
    Invitrogen™ iPrep™ Card: Viral DNA/RNA > 1 card