Learn More
Invitrogen™ iPrep™ Cartridge Rack
Stainless steel, alluminium alloy rack that holds the pre-filled carteridges during purification
Brand:  Invitrogen™ IS10102
Product Code. 10033013
  • £4085.00 / Each
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Description
Description
  • The iPrep™ Cartridge Rack is a stainless steel, alluminium alloy rack that holds the pre-filled carteridges during purification.
  • The rack is designed to fit onto the plaform in only one orientation (indicated with an arrow).
  • The racks dimensions are 10.8 inches (l) x 5.5 inches (w) x 2.3 inches (d).
  • Each iPrep Instrument includes 1 iPRep Carteridge Rack. Additional racks can be ordered for faster instrument set up.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Store at room temperature.
iPrep™
Room Temperature
iPrep™
1 rack (s)