Learn More
Iprovalicarb, Fluka™
Analytical standard, reference material
Brand: Honeywell Fluka R1641-100MG
Additional Details : CAS Number : 140923-17-7 Weight : 0.00010kg
Description
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
- Expert Technical Support
Specifications
Iprovalicarb | |
140923-17-7 | |
White | |
320.43 | |
164°C | |
Environmental Analysis |
Glass Bottle | |
100 mg | |
Isopropyl [(1S)-2-methyl-1-{[(1RS)-1-p-tolylethyl]carbamoyl}propyl]carbamate | |
Analytical Standard | |
Decomposes below the boiling point. |