Learn More
Thermo Scientific™ IPTG Solution, ready-to-use
Induce the expression of genes under the control of lac operon in cloning and protein expression procedures.
Brand: Thermo Scientific™ R1171
66.01 GBP valid until 2024-10-31
Use promo code "21744" to get your promotional price.
Description
IPTG (isopropyl-beta-D-thiogalactopyranoside) is a highly stable synthetic analog of lactose. It inactivates the lac repressor and induces synthesis of beta-galactosidase, an enzyme that promotes lactose utilization. IPTG is used to induce expression of cloned genes under control of the lac operon . It is used in conjunction with X-Gal (#R0941) to determine the lac phenotype in blue/white colony screening.
It inactivates the lac repressor and induces synthesis of beta-galactosidase, an enzyme that promotes lactose utilization. IPTG is used to induce expression of cloned genes under control of the lac operon. It is used in conjunction with X-Gal to determine the lac phenotype in blue/white colony screening.Ready-to-use 100mM IPTG aqueous solution is non-toxic, stable, 0.22μm membrane-filtered, formulated for direct use in blue/white colony screening.
Specifications
Isopropylthiogalactoside | |
Protein Expression | |
10 x 1.5 mL |
0.1 M | |
IPTG Solution | |
Liquid |
For Research Use Only. Not for use in diagnostic procedures.