Learn More
Thermo Scientific™ IPTG, dioxane-free
Induce the expression of genes under the control of lac operon in cloning and protein expression procedures.
£22.92 - £259.00
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10213090
View Documents
Thermo Scientific™
R0391
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10725471
View Documents
Thermo Scientific™
R0392
5 g
£64.50
5g
10849040
View Documents
Thermo Scientific™
R0393
5 x 5 g
£259.00
25g
Description
Description
IPTG (isopropyl-beta-D-thiogalactopyranoside) is a highly stable synthetic analog of lactose. It inactivates the lac repressor and induces synthesis of beta-galactosidase, an enzyme that promotes lactose utilization. IPTG is used to induce expression of cloned genes under control of the lac operon . It is used in conjunction with X-Gal (#R0941) to determine the lac phenotype in blue/white colony screening.
It inactivates the lac repressor and induces synthesis of beta-galactosidase, an enzyme that promotes lactose utilization. IPTG is used to induce expression of cloned genes under control of the lac operon. It is used in conjunction with X-Gal to determine the lac phenotype in blue/white colony screening.
Ready-to-use 100mM IPTG aqueous solution is non-toxic, stable, 0.22μm membrane-filtered, formulated for direct use in blue/white colony screening.
Videos
SDS
Documents
Documents
Product Certifications
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Product Title
Thermo Scientific™ IPTG, dioxane-free