Learn More
Memmert™ IQ/OQ Document with Device-Specific Works Test Data - Temperature and Humidity
For one free-selectable temperature and humidity value, Includes temperature distribution survey at Memmert to DIN 12880:2007-05
Brand:  Memmert™ HPP110ecoD00136
Additional Details : Weight : 0.01100kg
 View more versions of this product
Product Code. 17119684
  • £2755.00 / Each
Estimated Shipment: 29-08-2024
to see stock.
Description
Description
  • 27 measuring points (sizes 110 - 1060, 2200)
  • 26 measuring points (size 1400)
  • PQ checklist as support for validation by customer
Specifications
Specifications
HPP110eco
IQ/OQ Document with Device-specific Works Test Data