Learn More
Memmert™ IQ/OQ Document with Test Data for UF1060plus
IQ/OQ document with temperature distribution survey for one free-selectable temperature value.
Brand: Memmert™ UF1060plusD00127
Additional Details : Weight : 0.01200kg
Description
- Includes temperature distribution survey with 9 temperature measuring points (size 30) and 27 measuring points (sizes 55 - 1060) to DIN 12880:2007-05 (further temperature values on demand)
- PQ check list as support for validation by customer
Specifications
UF1060plus Oven | |
IQ/OQ Documentation | |
Test Data |