Learn More
Grant Instruments™ IQ/OQ Documentation
Offers equipment user greater comparability and verification with their instruments. Grant Instruments™ IQ/OQ Documentation simplifies installation and maintenance procedures, reduces common user errors, and builds a clear record of an instrument's performance throughout its life.
Brand: Grant Instruments™ IQOQ XUBA
This item is not returnable.
View return policy
Description
- Installation Qualification (IQ): instrument has been delivered and installed in accordance with manufacturer's requirements
- Operational Qualification (OQ): instrument is functioning in accordance with its specification
- Each IQ/OQ documentation set is specific to individual serial numbered equipment and is provided in an easy to use ring bound format.
- It is recommend to purchase IQ/OQ documentation at the time of ordering Grant Scientific equipment.
Specifications
All Grant XUBA Analogue Ultrasonic Baths | |
IQ/OQ Documentation |