Learn More
Implen™ IQ/OQ documentation for the NanoPhotometer™ C40 incl. Holmium Glass Filter and Neutral Density Glass Filter Set
In compliance with international standards in regulated environments, documented verification is provided with Installation Qualification and Operation Qualification (IQ/OQ) Package Conforming Data to verify that instruments are working according to specifications in case of an audit.
Brand: Implen™ N-40-QHN
Additional Details : Weight : 0.50000kg
Description
- Even though the Implen NanoPhotometer™ is considered recalibration-free, also non-regulated laboratories may utilize the IQOQ package, since it provides added peace of mind that the system is working appropriately and producing high-quality data.
- The NanoPhotomete™ IQOQ package consists of a liquid and/or a solid NIST traceable secondary standard and a corresponding Excel based software tool depending on the type of instrument.
- All secondary standards come with a certificate of compliance.
- The Excel based software produces an automated report displaying all relevant data for an audit.
Specifications
IQOQ for NanoPhotometer C40 | |
NanoPhotometer | |
Holmium and Neutral Density Glass Filter |
IQ/OQ Documentation | |
Wavescan |