Learn More
Implen™ IQ/OQ documentation for the NanoPhotometer™ NP80 incl. Holmium Glass Filter and Neutral Density Glass Filter Set and Standard Solution
In compliance with international standards in regulated environments, documented verification is provided with Installation Qualification and Operation Qualification (IQ/OQ) Package Conforming Data to verify that instruments are working according to specifications in case of an audit.
Brand: Implen™ N-80-QHN
Additional Details : Weight : 5.00000kg
Description
- Even though the Implen NanoPhotometer™ is considered recalibration-free, also non-regulated laboratories may utilize the IQOQ package, since it provides added peace of mind that the system is working appropriately and producing high-quality data.
- The NanoPhotomete™ IQOQ package consists of a liquid and/or a solid NIST traceable secondary standard and a corresponding Excel based software tool depending on the type of instrument.
- The liquid standard is based on a non-toxic compound absorbing at 280 nm to cover the area of most interest for Life Science Applications.
- All secondary standards come with a certificate of compliance.
- The Excel based software produces an automated report displaying all relevant data for an audit.
Specifications
IQOQ for NanoPhotometer NP80 | |
NanoPhotometer | |
Holmium and Neutral Density Glass Filter |
IQ/OQ Documentation | |
Wavescan |