Learn More
Irbesartan-d4, MedChemExpress
MedChemExpress Irbesartan D4 is the deuterium labeled Irbesartan, which is a highly potent and specific angiotensin II type 1 (AT1) receptor antagonist.
Brand: Medchem Express HY-B0202S
This item is not returnable.
View return policy
Description
Irbesartan D4 is the deuterium labeled Irbesartan, which is a highly potent and specific angiotensin II type 1 (AT1) receptor antagonist.Specifications
Irbesartan-d4 | |
1 mg | |
Research | |
SR-47436-d4 BMS-186295-d4 | |
432.55 | |
99.46% | |
Neuroscience-Neuromodulation |
1216883-23-6 | |
White | |
C25H24D4N6O | |
O=C1N(CC2=C([2H])C([2H])=C(C3=C(C4=NN=NN4)C=CC=C3)C([2H])=C2[2H])C(CCCC)=NC15CCCC5 | |
432.55 | |
Research | |
Solid |
Research purposes only