missing translation for 'onlineSavingsMsg'
Learn More
Iridium, 99.9%, (trace metal basis), powder, Thermo Scientific Chemicals Product Code.: 10713534

Iridium, 99.9%, (trace metal basis), powder, Thermo Scientific Chemicals

Product Code. 10713534
10 g, Glass bottle
Click to view available options
Quantity:
500 mg
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10713534

Brand: Thermo Scientific Acros 198090100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-88-5
Molecular Formula Ir
Molecular Weight (g/mol) 192.22
MDL Number MFCD00011062
InChI Key GKOZUEZYRPOHIO-UHFFFAOYSA-N
Synonym ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge
PubChem CID 23924
ChEBI CHEBI:49666
IUPAC Name iridium
SMILES [Ir]

Specifications

Color Gray
Assay Percent Range 99.9%
Quantity 10 g
Fieser 02,228; 09,257
Merck Index 15, 5129
Formula Weight 192.2
Grade Trace Metal Basis
Packaging Glass bottle
Form Amorphous Powder
Chemical Name or Material Iridium

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463372 Certificate TypeCertificate of Analysis Date06/06/2024 Catalog Number198090100,198095000
Lot NumberA0432403 Certificate TypeCertificate of Analysis Date15/01/2024 Catalog Number198090100,198095000
Lot NumberA0457543 Certificate TypeCertificate of Analysis Date03/11/2023 Catalog Number198090100,198095000
Lot NumberA0454535 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number198090100,198095000
Lot NumberA0394904 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number198090100,198095000
compliance-icons compliance-icons
Product Identifier
  • Iridium, powder
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 1
Hazard Statement
  • H319-Causes serious eye irritation.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iridium, 99.9%, (trace metal basis), powder, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.