missing translation for 'onlineSavingsMsg'
Learn More

Iridium(III) bromide hydrate, Ir 35.9% min

Product Code. p-7048961
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11356618

Brand: Thermo Scientific Alfa Aesar 012212.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 171769-74-7 | Br3Ir | 431.93 g/mol

Iridium(III) bromide hydrate is used as a catalyst in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 171769-74-7
Molecular Formula Br3Ir
Molecular Weight (g/mol) 431.93
MDL Number MFCD00049466
InChI Key HTFVQFACYFEXPR-UHFFFAOYSA-K
SMILES [Br-].[Br-].[Br-].[Ir+3]

Specifications

Melting Point 100°C (in -3H2O)
Physical Form Crystalline
Quantity 1 g
Solubility Information Soluble in water.
IUPAC Name iridium(3+) tribromide
Formula Weight 431.91 (Anhydrous)
Concentration or Composition (by Analyte or Components) Ir 35.9% min.
Chemical Name or Material Iridium(III) bromide hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.