missing translation for 'onlineSavingsMsg'
Learn More
Iridium(III) chloride, anhydrous, Ir 62% min, Thermo Scientific Chemicals Product Code.: 11346528

Iridium(III) chloride, anhydrous, Ir 62% min, Thermo Scientific Chemicals

Product Code. 11346528
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11346528

Brand: Thermo Scientific Alfa Aesar 012158.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Iridium(III) chloride is used in iridium electroplating, in making the standard meter bar of paris and crucibles. It is used for high temperature electrical contacts, as hardening agent, in tipping pens and compass bearings and Pt/Ir spark plugs. It is used for medical, surgical or other patient oriented applications, MMO (mixed metal oxide) titanium electrode coatings and as a starting material for heterogeneous catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-83-9
Molecular Formula Cl3Ir
Molecular Weight (g/mol) 298.57
MDL Number MFCD00011063
InChI Key DANYXEHCMQHDNX-UHFFFAOYSA-K
Synonym iridium iv chloride, iridium tetrachloride, iridium chloride ircl4, iridic chloride, unii-pcg7kvc21i, pcg7kvc21i, acmc-1cp8w, iridium iv chloride, premion
PubChem CID 24815
IUPAC Name tetrachloroiridium
SMILES [Cl-].[Cl-].[Cl-].[Ir+3]

Specifications

Melting Point 763°C (decomposition)
Color Black to Brown
Physical Form Powder
Quantity 1 g
Assay Percent Range 99.95% (Metal basis)
Solubility Information It is insoluble in water.
Formula Weight 298.56
Sensitivity Hygroscopic
Appearance Drak brown to black glassy amorphous pieces/powder
Chemical Name or Material Iridium(IV) chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iridium(III) chloride, anhydrous, Ir 62% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.