Learn More
Iridium(III) chloride hydrate, 99.8% (metals basis), Thermo Scientific Chemicals
Description
Iridium(III) chloride hydrate is used to prepare other iridium compounds. It finds use as a catalyst for oxidation in presence of cerium(IV) and in the the manufacture of electrodes. Iridium(III) chloride is used in iridium electroplating, in making the standard meter bar of Paris and crucibles. It is used for high temperature electrical contacts, as hardening agent, in tipping pens and compass bearings and Pt/Ir spark plugs. It is used for medical, surgical or other patient oriented applications, MMO (mixed metal oxide) titanium electrode coatings and as a starting material for heterogeneous catalysts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Black |
Physical Form | Crystals |
Quantity | 1 g |
Assay Percent Range | 99.9% (Metals basis) |
Solubility Information | Soluble in water and alcohol. |
Formula Weight | 298.56 (Anhydrous) |
Odor | Odorless |
Chemical Name or Material | Iridium(III) chloride hydrate |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
N07L074 | Certificate of Analysis | 10/02/2025 | 011030.14,011030.03,011030.06 |
Y26K049 | Certificate of Analysis | 06/12/2024 | 011030.03,011030.14,011030.06 |
X10K008 | Certificate of Analysis | 30/10/2024 | 011030.03,011030.14,011030.06 |
P13K040 | Certificate of Analysis | 25/03/2024 | 011030.03,011030.14,011030.06 |
Q11J024 | Certificate of Analysis | 09/05/2023 | 011030.03 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.