missing translation for 'onlineSavingsMsg'
Learn More

Iridium(III) chloride trihydrate, 53-56% Ir

Product Code. p-4509777 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 10216590

Brand: Thermo Scientific Acros 195500010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13569-57-8 | Cl3Ir·3H2O | 352.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13569-57-8
Molecular Formula Cl3Ir·3H2O
Molecular Weight (g/mol) 352.6
InChI Key LNJXVUXPFZKMNF-UHFFFAOYSA-K
Synonym iridium iii chloride trihydrate, trichloroiridium trihydrate, cl3ir.3h2o, acmc-1c5tf, iridium chloride ircl3 , trihydrate 7ci,8ci,9ci
PubChem CID 6102153
IUPAC Name trichloroiridium;trihydrate
SMILES O.O.O.Cl[Ir](Cl)Cl

Specifications

Melting Point 763.0°C
Color Black to Green
Physical Form Crystals
Quantity 1 g
Linear Formula IrCl3·3H2O
Fieser 01,132; 02,67
Merck Index 14, 5088
Solubility Information Solubility in water: insoluble in water. Other solubilities: insoluble in acids ans alkalies
Formula Weight 352.6
Percent Purity 53 to 56% Ir
Packaging Glass bottle
Specific Gravity 5.3
Density 5.3000g/mL
Chemical Name or Material Iridium(III) chloride trihydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.