missing translation for 'onlineSavingsMsg'
Learn More

Iridium(IV) chloride, 99.95% (metals basis), Ir 56.5 min

Product Code. p-4408127
Click to view available options
Quantity:
0.25 g
1 g
Unit Size:
0.25g
1g
This item is not returnable. View return policy

Product Code. 11386568

Brand: Thermo Scientific Alfa Aesar 012184.01

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10025-97-5 | Cl4Ir | 334.017 g/mol

Iridium(IV) chloride is used in chemical analysis as testing for nitric acid in the presence of nitrous acid, microscopy and in plating solutions. It acts as a catalyst precursor for the stereoselective reduction of cyclohexanones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-97-5
Molecular Formula Cl4Ir
Molecular Weight (g/mol) 334.017
MDL Number MFCD00016155
InChI Key CALMYRPSSNRCFD-UHFFFAOYSA-J
Synonym iridium iv chloride, iridium tetrachloride, iridium chloride ircl4, iridic chloride, unii-pcg7kvc21i, pcg7kvc21i, acmc-1cp8w, iridium iv chloride, premion
PubChem CID 24815
IUPAC Name tetrachloroiridium
SMILES Cl[Ir](Cl)(Cl)Cl

Specifications

Melting Point (decomposition)
Physical Form Glassy Amorphous Pieces, Powder
Quantity 0.25 g
Assay Percent Range 99.95% (Metal basis)
Solubility Information Soluble in water,alcohol and dilute hydrochloric acid.
Formula Weight 334.01
Sensitivity Hygroscopic
Appearance Drak brown to black glassy amorphous pieces/powder
Chemical Name or Material Iridium(IV) chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.