Learn More
Iridium powder, -325 mesh, 99.9% (metals basis), Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Physical Form | Powder, Sponge |
Quantity | 0.5 g |
UN Number | UN3089 |
Percent Purity | 99.9% |
Assay | (metals basis) |
Chemical Name or Material | Iridium powder |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Z09I008 | Certificate of Analysis | 05/01/2023 | 039684.04,039684.02 |
S23I045 | Certificate of Analysis | 30/06/2022 | 039684.04,039684.02 |
Q10G010 | Certificate of Analysis | 12/04/2022 | 039684.04,039684.02 |
R07G085 | Certificate of Analysis | 12/04/2022 | 039684.04,039684.02 |
E08U019 | Certificate of Analysis | 12/11/2021 | 039684.04,039684.02 |
Safety and Handling


- Iridium powder
- Danger
- Serious eye damage/eye irritation Category 2
- Flammable solid Category 1
- H228-Flammable solid.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Iridium
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.