missing translation for 'onlineSavingsMsg'
Learn More

Iridium shot, 6-7mm (0.24-0.28in), 99.95% (metals basis), Thermo Scientific Chemicals

Product Code. 15436555
1 Pc.
Click to view available options
Quantity:
1 Pc.
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7439-88-5
Molecular Formula Ir
MDL Number MFCD00011062
Quantity 1 Pc.
Synonym ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge
InChI Key GKOZUEZYRPOHIO-UHFFFAOYSA-N
SMILES [Ir]
IUPAC Name iridium
Molecular Weight (g/mol) 192.22
PubChem CID 23924
ChEBI CHEBI:49666
Percent Purity 99.95%
Weight ≈4.5g/piece
Form Pieces
Assay (metals basis)
Chemical Name or Material Iridium shot, 6 to 7mm (0.24 to 0.28 in.)
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iridium shot, 6-7mm (0.24-0.28in), 99.95% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.