missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iridium Standard For ICP, 1000μg/mL in 10% HCl, Assurance Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
£285.00 - £319.00
Chemical Identifiers
CAS | 7439-88-5, 7647-01-0, 7732-18-5 |
---|---|
Molecular Formula | Ir |
Molecular Weight (g/mol) | 192.22 |
MDL Number | MFCD00011062 |
InChI Key | GKOZUEZYRPOHIO-UHFFFAOYSA-N |
PubChem CID | 23924 |
ChEBI | CHEBI:49666 |
IUPAC Name | iridium |
SMILES | [Ir] |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10172772
|
SPEX Certiprep
PLIR3-2Y |
125 mL | Triple-leached LDPE bottle |
£285.00
125mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10365982
|
SPEX Certiprep
PLIR3-2X |
500 mL | Triple-leached LDPE bottle |
£319.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Ir | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
7439-88-5, 7647-01-0, 7732-18-5 | |
192.22 | |
GKOZUEZYRPOHIO-UHFFFAOYSA-N | |
CHEBI:49666 | |
[Ir] |
Ir | |
MFCD00011062 | |
23924 | |
iridium |
Specifications
7439-88-5, 7647-01-0, 7732-18-5 | |
0.1, 10.0, 89.9 | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Ir | |
GKOZUEZYRPOHIO-UHFFFAOYSA-N | |
iridium | |
23924 | |
Plasma | |
10% HCl |
0.1, 10.0, 89.9 | |
Assurance Certificate of Analysis reports up to 39 elements at ppb levels. | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
MFCD00011062 | |
[Ir] | |
192.22 | |
CHEBI:49666 | |
Matrix Concentration Cat. No. 10% HCl 1000μg/mL PLIR3-2Y, -2X |
|
Iridium Standard |
Safety and Handling
missing translation for 'dotInformation' : Hazardous
Recommended Storage : Ambient
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 1000
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P405-Store locked up.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : hydrochloric acid, Iridium from Iridium(III) chloride hydrate
Product intended for laboratory use only