missing translation for 'onlineSavingsMsg'
Learn More
Iridium wire, 1.0mm (0.04in) dia, 99.8% (metals basis), Thermo Scientific Chemicals Product Code.: 11351259

Iridium wire, 1.0mm (0.04in) dia, 99.8% (metals basis), Thermo Scientific Chemicals

Product Code. 11351259
5 cm
Click to view available options
Quantity:
1 cm
5 cm
25 cm
This item is not returnable. View return policy

Product Code. 11351259

Brand: Thermo Scientific Alfa Aesar 011429.BQ

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-88-5
Molecular Formula Ir
Molecular Weight (g/mol) 192.22
MDL Number MFCD00011062
InChI Key GKOZUEZYRPOHIO-UHFFFAOYSA-N
Synonym ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge
PubChem CID 23924
ChEBI CHEBI:49666
IUPAC Name iridium
SMILES [Ir]

Specifications

Quantity 5 cm
Percent Purity 99.8%
Weight ≈17.8g/m
Form Wire
Assay (metals basis)
Chemical Name or Material Iridium wire, 1.0mm (0.04 in.) dia.

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberH13U001 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number011429.BQ,011429.BO,011432.BS
Lot NumberB08Z032 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number011429.BQ,011429.BO,011432.BS
Lot NumberG03Y003 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number011429.BQ,011429.BO,011432.BS
Lot NumberZ17A008 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number011429.BQ,011429.BO,011432.BS
Lot NumberE18U010 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number011429.BQ,011429.BO,011432.BS

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iridium wire, 1.0mm (0.04in) dia, 99.8% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.