missing translation for 'onlineSavingsMsg'
Learn More

Iron-chrome based high temperature gas shift catalyst, HiFUEL∣r W210, Thermo Scientific Chemicals

Product Code. 15456935
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Iron-chrome based high temperature gas shift catalyst is ideal for use in fuel cell and other distributed hydrogen production applications. It is also used for fuel processing applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Color Red to Brown
Quantity 500 g
Solubility Information Insoluble in water.
Physical Form Pellets
Chemical Name or Material Iron-chrome based high temperature gas shift catalyst HiFUEL™ W210
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Iron-chrome based high temperature gas shift catalyst, HiFUEL W213
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
  • Germ cell mutagenicity Category 1A
  • Carcinogenicity Category 1A
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H332-Harmful if inhaled.
  • H317-May cause an allergic skin reaction.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : Chromium (III) oxide, Copper (II) oxide,Chromium (VI) oxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'