missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron granules, 1-2mm (0.04-0.08in), 99.98% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Gray |
Appearance | Gray color granules |
Density | 7.87g/cm3 at 20°C |
Assay Percent Range | 99.98% (Metals basis) |
Quantity | 250 g |
Form | Granular |
Chemical Name or Material | Iron |
Safety and Handling

Product Identifier
- Iron granules
- Warning
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Iron granules, 1-2mm (0.04-0.08in), 99.98% (metals basis), Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction