missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(II) 2-ethylhexanoate monoisopropoxide, 5% w/v in hexane, Thermo Scientific™
Click to view available options
Quantity:
25 mL
100 mL
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Quantity | 25 mL |
UN Number | UN1208 |
Sensitivity | Moisture sensitive |
Formula Weight | 258.14 |
Concentration or Composition (by Analyte or Components) | 5% w/v in hexane |
Physical Form | Liquid |
Chemical Name or Material | Iron(II) 2-ethylhexanoate monoisopropoxide |
Safety and Handling
Product Identifier
- Iron(II) 2-ethylhexanoate monoisopropoxide, 5% w/v in hexane
- Danger
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
- Aspiration toxicity Category 1
- LONG-TERM AQUATIC HAZARD Chronic 2
- H225-Highly flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H361f-Suspected of damaging fertility.
- H336-May cause drowsiness or dizziness.
- H373-May cause damage to organs.
- H304-May be fatal if swallowed and enters airways.
- H411-Toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P273-Avoid release to the environment.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P315-Get immediate medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction