missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) bromide, 98%, anhydrous, Thermo Scientific Chemicals

Product Code. 10649643
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-46-0
Molecular Formula Br2Fe
Molecular Weight (g/mol) 215.65
MDL Number MFCD00016081
InChI Key GYCHYNMREWYSKH-UHFFFAOYSA-L
Synonym iron dibromide, unii-ea3x054rbz, iron ii bromide, ea3x054rbz, iron 2+ dibromide, iron ion 2+ dibromide, iron ii bromide, anhydrous 5g, iron ii bromide, ultra dry metals basis, ferrous bromide, anhydrous
PubChem CID 82240
SMILES [Fe++].[Br-].[Br-]

Specifications

Boiling Point 937°C
Melting Point 684°C
Color Black to Brown-Red
Physical Form Crystals or Powder
Quantity 10 g
Assay Percent Range 97.5% min. (Argentometry)
Linear Formula FeBr2
Merck Index 15, 4069
Solubility Information Solubility in water: 1090g/L
IUPAC Name λ²-iron(2+) dibromide
Formula Weight 215.65
Percent Purity 98%
Packaging Glass bottle
Chemical Name or Material Iron(II) bromide
Show More Show Less
compliance-icons
Identifiant produit
  • Iron(II) bromide
Mention d'avertissement
  • Danger
Catégorie de danger
  • Skin corrosion/irritation Category 1 C
  • Serious eye damage/eye irritation Category 1
Mention de danger
  • H314-Causes severe skin burns and eye damage.
Conseils de prudence
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'