Learn More
Iron(II) chloride, 97%, anhydrous, Thermo Scientific Chemicals
£98.79 - £541.70
Chemical Identifiers
CAS | 7758-94-3 |
---|---|
Molecular Formula | Cl2Fe |
Molecular Weight (g/mol) | 126.75 |
MDL Number | MFCD00011004 |
InChI Key | NMCUIPGRVMDVDB-UHFFFAOYSA-L |
Synonym | Ferrous chloride |
IUPAC Name | λ²-iron(2+) dichloride |
SMILES | [Cl-].[Cl-].[Fe++] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11904191
|
Thermo Scientific Acros
389350250 |
25 g | Glass bottle |
£98.79
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13418538
|
Thermo Scientific Acros
389352500 |
250 g | Glass bottle |
£541.70
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7758-94-3 | |
126.75 | |
NMCUIPGRVMDVDB-UHFFFAOYSA-L | |
λ²-iron(2+) dichloride |
Cl2Fe | |
MFCD00011004 | |
Ferrous chloride | |
[Cl-].[Cl-].[Fe++] |
Specifications
1026.0°C | |
7758-94-3 | |
Powder | |
Cl2Fe | |
MFCD00011004 | |
Ferrous chloride | |
NMCUIPGRVMDVDB-UHFFFAOYSA-L | |
λ²-iron(2+) dichloride | |
126.75 | |
Iron(II) chloride |
677.0°C | |
Brown | |
97% | |
FeCl2 | |
15,407 | |
Solubility in water: 644g/L | |
[Cl-].[Cl-].[Fe++] | |
126.75 | |
97% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 231-843-4
- Iron(II) chloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 C
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.