missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) chloride tetrahydrate, 99+%

Product Code. 10401251 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic bottle
missing translation for 'unitSize'
1kg
250g
5g
5kg
This item is not returnable. View return policy

Product Code. 10401251

missing translation for 'mfr': Thermo Scientific Acros 205082500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 13478-10-9 | Cl2Fe·4H2O | 198.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13478-10-9
Molecular Formula Cl2Fe·4H2O
Molecular Weight (g/mol) 198.81
InChI Key WSSMOXHYUFMBLS-UHFFFAOYSA-L
Synonym ferrous chloride tetrahydrate, iron ii chloride tetrahydrate, iron dichloride tetrahydrate, cl2fe.4h2o, fecl2.4h2o, iron chloride, tetrahydrate, dichloroiron-water 1/4, iron ii chloride tetra hydrate, ae>> nciu eae r masculineiii
PubChem CID 16211588
ChEBI CHEBI:86249
IUPAC Name dichloroiron;tetrahydrate
SMILES O.O.O.O.Cl[Fe]Cl

Specifications

Melting Point 105.0°C to 110.0°C
Color Yellow to Green
Physical Form Crystalline Powder, Crystals and/or Chunks
Quantity 250 g
Assay Percent Range 99+%
Linear Formula FeCl2·4H2O
Merck Index 14,4043
Solubility Information Solubility in water: 1600g/L (10°C) 4100g/L (100°C). Other solubilities: soluble in alcohol
Formula Weight 198.81
Percent Purity ≥99%
Packaging Glass bottle
Chemical Name or Material Iron(II) chloride tetrahydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iron(II) chloride tetrahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.