missing translation for 'onlineSavingsMsg'
Learn More

Iron(II,III) oxide, nanopowder, 97%, Thermo Scientific Chemicals

£29.35 - £407.03

Chemical Identifiers

CAS 1317-61-9
Molecular Formula Fe3O4
Molecular Weight (g/mol) 231.53
MDL Number MFCD00011010
InChI Key WTFXARWRTYJXII-UHFFFAOYSA-N
IUPAC Name diiron(3+) λ²-iron(2+) tetraoxidandiide
SMILES [O--].[O--].[O--].[O--].[Fe++].[Fe+3].[Fe+3]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15432867
View Documents
Thermo Scientific Alfa Aesar
047141.14
25 g
£29.35
25g
Please to purchase this item. Need a web account? Register with us today!
15442867
View Documents
Thermo Scientific Alfa Aesar
047141.22
100 g
£94.57
100g
Please to purchase this item. Need a web account? Register with us today!
15452867
View Documents
Thermo Scientific Alfa Aesar
047141.36
500 g
£407.03
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Iron(II,III) oxide is mainly used as a black-pigment-catalyst in the Haber process and in the water gas shift reaction. It is used as a contrast-agent in MRI scanning and as a pigment in magnetic applications, polishing compounds, cosmetics, medicines, polymers, rubber, fillers, building, construction, appliances and magnetic inks.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1317-61-9
231.53
WTFXARWRTYJXII-UHFFFAOYSA-N
[O--].[O--].[O--].[O--].[Fe++].[Fe+3].[Fe+3]
Fe3O4
MFCD00011010
diiron(3+) λ²-iron(2+) tetraoxidandiide

Specifications

1538°C
Black
Fe3O4
14,4040
WTFXARWRTYJXII-UHFFFAOYSA-N
diiron(3+) λ²-iron(2+) tetraoxidandiide
231.54
Odorless
5.2 g/mL
1317-61-9
Powder
MFCD00011010
Insoluble in water and organic solvents. Soluble in concentrated mineral acids.
[O--].[O--].[O--].[O--].[Fe++].[Fe+3].[Fe+3]
231.53
97%
3.0
Iron(II,III) oxide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

missing translation for 'einecsNumber' : 215-277-5

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Iron(II,III) oxide, nanopowder

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only