missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(II) iodide, anhydrous, 97% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
2 g
10 g
Description
Iron(II) iodide is used as a catalyst in organic reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Brown |
Physical Form | Crystalline |
Quantity | 2 g |
UN Number | UN3260 |
Solubility Information | Soluble in water,diethyl ether and ethanol. |
Formula Weight | 309.66 |
Percent Purity | 97% |
Odor | Odorless |
Sensitivity | Air and light sensitive; Hygroscopic |
Density | 5.32 g/mL |
Show More |
Safety and Handling
Product Identifier
- Iron(II) iodide, anhydrous
- Danger
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 C
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'