Learn More
Iron(II) oxide, 99.5% (metals basis), Thermo Scientific Chemicals
In manufacture of green, heat-absorbing glass
£136.00 - £542.00
Chemical Identifiers
CAS | 1345-25-1 |
---|---|
Molecular Formula | FeO |
Molecular Weight (g/mol) | 71.844 |
MDL Number | MFCD00016095 |
InChI Key | UQSXHKLRYXJYBZ-UHFFFAOYSA-N |
Synonym | iron ii oxide, ferrous oxide, iron monooxide, iron monoxide, iron oxide, wustite, natural wuestite, eisenoxydul, hsdb 464, eisen ii-oxid |
PubChem CID | 14945 |
ChEBI | CHEBI:50820 |
IUPAC Name | oxoiron |
SMILES | O=[Fe] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11367288
|
Thermo Scientific Alfa Aesar
030513.06 |
5 g |
£136.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11357288
|
Thermo Scientific Alfa Aesar
030513.14 |
25 g |
£542.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
In manufacture of green, heat-absorbing glass; in steel manufacture; and as a catalystIron(II) oxide is used in manufacturing of green, heat-absorbing glass; in steel manufacture; and as a catalyst. It is used as a dye or pigment in pottery, glazes and glasses. It finds application as heat-absorbing glass used in buildings and automobiles. It is used as a precursor in the production of steel. It is also used in cosmetics and some tattoo inks. It can also be used for filtering phosphates from home aquaria and acts as a catalyst in a number of industrial and chemical operations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1345-25-1 | |
71.844 | |
UQSXHKLRYXJYBZ-UHFFFAOYSA-N | |
14945 | |
oxoiron |
FeO | |
MFCD00016095 | |
iron ii oxide, ferrous oxide, iron monooxide, iron monoxide, iron oxide, wustite, natural wuestite, eisenoxydul, hsdb 464, eisen ii-oxid | |
CHEBI:50820 | |
O=[Fe] |
Specifications
1369°C | |
99.5% (metals basis) | |
MFCD00016095 | |
Insoluble in water and alkalies. Soluble in HCl,H2SO4. Slightly soluble in HNO3Insoluble in water,alcohol and alkali. Soluble in hydrochloric acid and sulfuric acid. Slightly soluble in nitric acid. | |
O=[Fe] | |
71.844 | |
CHEBI:50820 | |
Air Sensitive | |
Iron(II) oxide |
1345-25-1 | |
FeO | |
iron ii oxide, ferrous oxide, iron monooxide, iron monoxide, iron oxide, wustite, natural wuestite, eisenoxydul, hsdb 464, eisen ii-oxid | |
UQSXHKLRYXJYBZ-UHFFFAOYSA-N | |
oxoiron | |
14945 | |
71.85 | |
5.7 |
Safety and Handling
missing translation for 'einecsNumber' : 215-721-8
missing translation for 'tsca' : Yes
missing translation for 'storageNote' : Air Sensitive
Recommended Storage : Ambient temperatures
RUO – Research Use Only