missing translation for 'onlineSavingsMsg'
Learn More
Iron(II) sulfate heptahydrate, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, 99.5-104.5% (manganometric), Honeywell Fluka™ Product Code.: 15661140

Iron(II) sulfate heptahydrate, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, 99.5-104.5% (manganometric), Honeywell Fluka™
BIOPHARMA

Product Code. 15661140 Shop All Honeywell Fluka Products
500 g
Click to view available options
Quantity:
250 g
500 g
1 kg
2.5 kg
5 kg
25 kg
4 x 5 kg
6 x 1 kg
This item is not returnable. View return policy

Product Code. 15661140

Brand: Honeywell-Fluka 12354500G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP, 99.5-104.5% (manganometric)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-63-0
Molecular Formula FeH14O11S
Molecular Weight (g/mol) 278.01
MDL Number MFCD00149719
InChI Key SURQXAFEQWPFPV-UHFFFAOYSA-L
Synonym iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate
PubChem CID 62662
ChEBI CHEBI:75836
IUPAC Name iron(2+);sulfate;heptahydrate
SMILES O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O

Specifications

Quantity 500 g
Linear Formula FeSO4 · 7H2O
Formula Weight 278.01g/mol
Percent Purity 99.5 to 104.5% (manganometric)
Grade puriss.
Packaging Plastic Bottle
Vapor Pressure 14.6mmHg (25°C)
Density 1.898g/mL at 25°C
Chemical Name or Material Iron(Ii) Sulfate Heptahydrate, Meets analytical specification of Ph. Eur.BP, USP, FCC
compliance-icons
Product Identifier
  • Iron(II) sulfate heptahydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Iron(II) sulfate-7-hydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iron(II) sulfate heptahydrate, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, 99.5-104.5% (manganometric), Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.