Learn More
Iron(III) 2-ethylhexanoate, nominally 50% in mineral spirits, Fe 6%, Thermo Scientific™
£22.60 - £159.00
Chemical Identifiers
CAS | 7321-53-1 |
---|---|
Molecular Formula | C24H45FeO6 |
Molecular Weight (g/mol) | 485.463 |
MDL Number | MFCD00014006 |
InChI Key | ULUYRVWYCIOFRV-UHFFFAOYSA-K |
Synonym | ferric 2-ethylhexanoate, iron 2-ethylhexanoate, iron tris 2-ethylhexanoate, iron ethylhexanoate, iron iii 2-ethylhexanoate, hexanoic acid, 2-ethyl-, iron 3+ salt 3:1, iron iii 2-ethylhexanoate nominally in mineral spirits, fe, iron tallate 2-ethylhexanoate, iron 3+ tris 2-ethylhexanoate, hexanoic acid, 2-ethyl-, iron 3+ salt |
PubChem CID | 110963 |
IUPAC Name | 2-ethylhexanoate;iron(3+) |
SMILES | CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Fe+3] |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
7321-53-1 | |
485.463 | |
ULUYRVWYCIOFRV-UHFFFAOYSA-K | |
110963 | |
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Fe+3] |
C24H45FeO6 | |
MFCD00014006 | |
ferric 2-ethylhexanoate, iron 2-ethylhexanoate, iron tris 2-ethylhexanoate, iron ethylhexanoate, iron iii 2-ethylhexanoate, hexanoic acid, 2-ethyl-, iron 3+ salt 3:1, iron iii 2-ethylhexanoate nominally in mineral spirits, fe, iron tallate 2-ethylhexanoate, iron 3+ tris 2-ethylhexanoate, hexanoic acid, 2-ethyl-, iron 3+ salt | |
2-ethylhexanoate;iron(3+) |
Specifications
7321-53-1 | |
0.91 g/mL | |
MFCD00014006 | |
ferric 2-ethylhexanoate, iron 2-ethylhexanoate, iron tris 2-ethylhexanoate, iron ethylhexanoate, iron iii 2-ethylhexanoate, hexanoic acid, 2-ethyl-, iron 3+ salt 3:1, iron iii 2-ethylhexanoate nominally in mineral spirits, fe, iron tallate 2-ethylhexanoate, iron 3+ tris 2-ethylhexanoate, hexanoic acid, 2-ethyl-, iron 3+ salt | |
ULUYRVWYCIOFRV-UHFFFAOYSA-K | |
2-ethylhexanoate;iron(3+) | |
110963 | |
Fe 6% | |
Iron (III) 2-ethylhexanoate, Nominally 50% in mineral spirits |
Brown | |
C24H45FeO6 | |
UN1268 | |
Miscible with chloroform. Immiscible with water. | |
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Fe+3] | |
485.463 | |
485.47 | |
Liquid |
Safety and Handling
GHS H Statement
H340-H350-H372-H304-H226
May cause genetic defects.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
P201-P202-P210-P233-P240-P241-P242-P243-P260-P264b-P270-P281-P301+P310-P303+P361+P353-P308+P313-P331-P370+P378q-P501c
H226-H304-H340-H350-H372
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PETROLEUM DISTILLATES, N.O.S.
missing translation for 'einecsNumber' : 230-794-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Iron(III) 2-ethylhexanoate, nominally 50% in mineral spirits, Fe 6%
Signal Word
- Danger
Hazard Category
- Germ cell mutagenicity Category 1B
- Carcinogenicity Category 1B
- Specific target organ toxicity Category 1
- Aspiration toxicity Category 1
Hazard Statement
- H340-May cause genetic defects.
- H350-May cause cancer.
- H372-Causes damage to organs.
- H304-May be fatal if swallowed and enters airways.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P405-Store locked up.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : Mineral spirits