missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(III) acetylacetonate, 99+%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
250 g
1 kg
Packaging:
Glass bottle
missing translation for 'unitSize'
100g
1kg
250g
25g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 180.0°C to 183.0°C |
| Color | Red |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99% min. (Iodometry) |
| Packaging | Glass bottle |
| Quantity | 25 g |
| Fieser | 04,268; 06,304; 13,211 |
| Solubility Information | Solubility in water: 2g/L (20°C). Other solubilities: readily soluble in acetone and chloroform,sparingly soluble in ether,40.1g/kg alcohol (20°C),535.4g/kg benzene (20°C) |
| Formula Weight | 353.17 |
| Percent Purity | 99+% |
| Show More |
Safety and Handling
Product Identifier
- Iron(III) acetylacetonate
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction