missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) acetylacetonate, 99+%, Thermo Scientific Chemicals

Product Code. 15169173
Click to view available options
Quantity:
25 g
100 g
250 g
1 kg
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14024-18-1
Molecular Formula C15H21FeO6
Molecular Weight (g/mol) 353.17
MDL Number MFCD00000020
InChI Key AQBLLJNPHDIAPN-LNTINUHCSA-K
Synonym 2, 4-Pentanedione, iron(III) derivative, Ferric acetylacetonate
IUPAC Name iron(3+) tris((2Z)-4-oxopent-2-en-2-olate)
SMILES [Fe+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 180.0°C to 183.0°C
Color Red
Infrared Spectrum Authentic
Assay Percent Range 99% min. (Iodometry)
Packaging Glass bottle
Quantity 1 kg
Fieser 04,268; 06,304; 13,211
Solubility Information Solubility in water: 2g/L (20°C). Other solubilities: readily soluble in acetone and chloroform,sparingly soluble in ether,40.1g/kg alcohol (20°C),535.4g/kg benzene (20°C)
Formula Weight 353.17
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material Iron(III) acetylacetonate, 99+%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'