missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(III) Chloride Anhydrous, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
250 g
500 g
3 kg
Description
Iron(III) Chloride Anhydrous, Extra Pure, SLR
Specifications
Specifications
Color | Gray |
pH | 2 |
Physical Form | Powder/Solid |
Quantity | 3 kg |
Formula Weight | 162.21g/mol |
Packaging | Plastic powder jar |
Vapor Pressure | 1 hPa at 20°C |
Safety and Handling
Product Identifier
- Iron (III) chloride anhydrous
- Danger
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- Skin sensitiser Category 1
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- EUH208-Contains . May produce an allergic reaction.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'