missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) Chloride Anhydrous, Extra Pure, SLR, Fisher Chemical™

Product Code. 10224390 Shop All Fisher Chemical Products
Click to view available options
Quantity:
250 g
500 g
3 kg
Unit Size:
250g
3kg
500g
This item is not returnable. View return policy

Product Code. 10224390

Brand: Fisher Chemical I/1035/53

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Iron(III) Chloride Anhydrous, Extra Pure, SLR

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7705-08-0
Molecular Formula Cl3Fe
Molecular Weight (g/mol) 162.20
MDL Number MFCD00011005
InChI Key RBTARNINKXHZNM-UHFFFAOYSA-K
IUPAC Name iron(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Color Gray
pH 2
Physical Form Powder/Solid
Quantity 500 g
Formula Weight 162.21g/mol
Packaging Plastic powder jar
Vapor Pressure 1 hPa at 20°C
compliance-icons compliance-icons
Product Identifier
  • Iron (III) chloride anhydrous
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.