missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(III) Chloride Anhydrous, Extra Pure, SLR, Fisher Chemical™
£47.35 - £204.00
Chemical Identifiers
CAS | 7705-08-0 |
---|---|
Molecular Formula | Cl3Fe |
Molecular Weight (g/mol) | 162.20 |
MDL Number | MFCD00011005 |
InChI Key | RBTARNINKXHZNM-UHFFFAOYSA-K |
IUPAC Name | iron(3+) trichloride |
SMILES | [Cl-].[Cl-].[Cl-].[Fe+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10407260
|
Fisher Chemical
I/1035/50 |
250 g |
£47.35
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10224390
|
Fisher Chemical
I/1035/53 |
500 g |
£62.55
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10050170
|
Fisher Chemical
I/1035/63 |
3 kg |
£204.00
3kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Iron(III) Chloride Anhydrous, Extra Pure, SLR
Chemical Identifiers
7705-08-0 | |
162.20 | |
RBTARNINKXHZNM-UHFFFAOYSA-K | |
[Cl-].[Cl-].[Cl-].[Fe+3] |
Cl3Fe | |
MFCD00011005 | |
iron(3+) trichloride |
Specifications
7705-08-0 | |
2 | |
Cl3Fe | |
RBTARNINKXHZNM-UHFFFAOYSA-K | |
iron(3+) trichloride | |
162.21g/mol | |
1 hPa at 20°C |
Gray | |
Powder/Solid | |
MFCD00011005 | |
[Cl-].[Cl-].[Cl-].[Fe+3] | |
162.20 | |
Plastic powder jar |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Iron (III) chloride anhydrous
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.